missing translation for 'onlineSavingsMsg'
Learn More

7-Aminothieno[2,3-b]pyrazine-6-carboxylic acid, ≥90%, Thermo Scientific™

Product Code. p-3174723
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10387354 250 mg Amber Glass Bottle 250mg
10641654 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10387354 Supplier Thermo Scientific Maybridge Supplier No. MO07475CB
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 56881-31-3
Molecular Formula C7H5N3O2S
Molecular Weight (g/mol) 195.196
MDL Number MFCD08271964
InChI Key UJUCBOIXAMPUQL-UHFFFAOYSA-N
Synonym 7-aminothieno 2,3-b pyrazine-6-carboxylic acid, 7-aminothieno 2,3-b pyrazine-6-carboxylicacid, 7-aminothiopheno 3,2-b pyrazine-6-carboxylic acid, thieno 2,3-b pyrazine-6-carboxylicacid, 7-amino, 7-aminothieno 2,3-b pyrazine-6-carboxylic acid mb41-1
PubChem CID 12225712
IUPAC Name 7-aminothieno[2,3-b]pyrazine-6-carboxylic acid
SMILES C1=CN=C2C(=N1)C(=C(S2)C(=O)O)N

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 195.2
Percent Purity 90%
Chemical Name or Material 7-Aminothieno[2, 3-b]pyrazine-6-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.