missing translation for 'onlineSavingsMsg'
Learn More

(7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate, 99+%

Product Code. 15495265
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15495265 5 g 5g
15485265 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495265 Supplier Thermo Scientific Alfa Aesar Supplier No. H54378.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 156311-83-0 | C17H27F6N7OP2 | 521.39 g/mol

(7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate is used as reagent for synthesis of cyclic RGC pentapeptides for functionalization through click chemistry, fluorescent glucose bioprobes and ReactIR™ flow cell. It is also used as reagent for disulfide bond engineering, heteroaryl ether synthesis and optimization of conformational constraint on non-phosphorylated cyclic peptide antagonists of the Grb2-SH2 domain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate is used as reagent for synthesis of cyclic RGC pentapeptides for functionalization through click chemistry, fluorescent glucose bioprobes and ReactIR™ flow cell. It is also used as reagent for disulfide bond engineering, heteroaryl ether synthesis and optimization of conformational constraint on non-phosphorylated cyclic peptide antagonists of the Grb2-SH2 domain.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156311-83-0
Molecular Formula C17H27F6N7OP2
Molecular Weight (g/mol) 521.39
MDL Number MFCD03703417
InChI Key CBZAHNDHLWAZQC-UHFFFAOYSA-N
Synonym pyaop, 3h-1,2,3 triazolo 4,5-b pyridin-3-yl oxy tri pyrrolidin-1-yl phosphonium hexafluorophosphate v, 7-azabenzotriazol-1-yloxy tripyrrolidinophosphonium hexafluorophosphate, 3-hydroxy-3h-1,2,3-triazolo 4,5-b pyridinato-o tri-1-pyrrolidinylphosphonium hexafluorophosphate, pyaop reagent, 7-azabenzotriazol-1-yloxy tripyrrolidino-phosphonium hexafluorophosphate, 7-azabenzotriazol-1-yloxy-tris-pyrrolidino phosphonium hexafluorophosphate, pubchem12746, ksc491e1h
PubChem CID 11038641
IUPAC Name tripyrrolidin-1-yl(triazolo[4,5-b]pyridin-3-yloxy)phosphanium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.C1CCN(C1)[P+](ON1N=NC2=CC=CN=C12)(N1CCCC1)N1CCCC1

Specifications

Melting Point 163°C to 168°C
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 521.38
Percent Purity ≥99%
Chemical Name or Material (7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.