missing translation for 'onlineSavingsMsg'
Learn More

7-Azaindole-3-carboxylic acid, 95%

Product Code. 15456298
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15456298 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15456298 Supplier Thermo Scientific Alfa Aesar Supplier No. H59866.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 156270-06-3 | C8H6N2O2 | 162.148 g/mol

7-Azaindole-3-carboxylic acid is a reactant for synthesis of azaindol derivatives as new acrosin inhibitors and for preparation of triazoles via regioselective heterocyclizaiton reactions. It is also a reactant for synthesis of azaindolylcarboxy-endo-tropanamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Azaindole-3-carboxylic acid is a reactant for synthesis of azaindol derivatives as new acrosin inhibitors and for preparation of triazoles via regioselective heterocyclizaiton reactions. It is also a reactant for synthesis of azai
TRUSTED_SUSTAINABILITY

Specifications

CAS 156270-06-3
Melting Point 205°C to 209°C
Molecular Formula C8H6N2O2
MDL Number MFCD07778360
Quantity 1 g
Synonym 1h-pyrrolo 2,3-b pyridine-3-carboxylic acid, 7-azaindole-3-carboxylic acid, 1h-pyrrolo 2,3-b pyridine-3-carboxylicacid, pubchem20644, acmc-209zs1, ksc173s3p, 7-azaindole-3-carboxylic ..., methyl 7-azaindole-3-carboxylic acid
Solubility Information Slightly soluble in water.
InChI Key KYBIRFFGAIFLPM-UHFFFAOYSA-N
SMILES C1=CC2=C(NC=C2C(=O)O)N=C1
IUPAC Name 1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid
Molecular Weight (g/mol) 162.148
PubChem CID 10154191
Formula Weight 162.15
Percent Purity 95%
Chemical Name or Material 7-Azaindole-3-carboxylic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.