missing translation for 'onlineSavingsMsg'
Learn More

7-Azaindole-5-boronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 15575166 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15575166 1 g 1g
15585166 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15575166 Supplier Thermo Scientific Acros Supplier No. 455850010
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 754214-56-7
Molecular Formula C13H17BN2O2
Molecular Weight (g/mol) 244.10
MDL Number MFCD08060937
InChI Key UOXAMYZTYZLSCC-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 7-azaindole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, pyrrolo 2,3-b pyridine-5-boronic acid, pinacol ester, pyrrolo 2,3-b pyridin-5-ylboronic acid pinacol ester, zlchem 1246, pubchem16626, acmc-209szp, ksc641k4p
PubChem CID 24208789
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine
SMILES CC1(C)OB(OC1(C)C)C1=CN=C2NC=CC2=C1

Specifications

Melting Point 243.0°C to 245.5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 244.1
Percent Purity 97%
Chemical Name or Material 7-Azaindole-5-boronic acid pinacol ester
compliance-icons
Product Identifier
  • 7-Azaindole-5-boronic acid, pinacol ester, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.