missing translation for 'onlineSavingsMsg'
Learn More

7-Azaindole-5-boronic acid pinacol ester, 97%

Product Code. 11454430
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454430 250 mg 250mg
11464430 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454430 Supplier Thermo Scientific Alfa Aesar Supplier No. H50044.MD
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 754214-56-7 | C13H17BN2O2 | 244.10 g/mol

7-Azaindole-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Azaindole-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Soluble in methanol, ethanol and THF.

Notes
Air sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 754214-56-7
Molecular Formula C13H17BN2O2
Molecular Weight (g/mol) 244.10
MDL Number MFCD08060937
InChI Key UOXAMYZTYZLSCC-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 7-azaindole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, pyrrolo 2,3-b pyridine-5-boronic acid, pinacol ester, pyrrolo 2,3-b pyridin-5-ylboronic acid pinacol ester, zlchem 1246, pubchem16626, acmc-209szp, ksc641k4p
PubChem CID 24208789
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine
SMILES CC1(C)OB(OC1(C)C)C1=CN=C2NC=CC2=C1

Specifications

Melting Point 241°C to 245°C
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Soluble in methanol,ethanol and THF.
Formula Weight 244.1
Percent Purity 97%
Chemical Name or Material 7-Azaindole-5-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.