missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-1,2,3,4-tetrahydroisoquinoline, 97%

Product Code. 11309929
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309929 1 g 1g
11319929 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309929 Supplier Thermo Scientific Alfa Aesar Supplier No. B25712.03
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 17680-55-6 | C9H10BrN | 212.09 g/mol

7-Bromo-1,2,3,4-tetrahydroisoquinoline is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Bromo-1,2,3,4-tetrahydroisoquinoline is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17680-55-6
Molecular Formula C9H10BrN
Molecular Weight (g/mol) 212.09
MDL Number MFCD06739047
InChI Key OYODEQFZAJVROF-UHFFFAOYSA-N
Synonym 7-bromo-1,2,3,4-tetrahydro-isoquinoline, isoquinoline, 7-bromo-1,2,3,4-tetrahydro, acmc-209uo4, 7-bromotetrahydroisoquinoline
PubChem CID 10729255
IUPAC Name 7-bromo-1,2,3,4-tetrahydroisoquinoline
SMILES C1CNCC2=C1C=CC(=C2)Br

Specifications

Melting Point 32°C to 35°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 212.09
Percent Purity 97%
Chemical Name or Material 7-Bromo-1,2,3,4-tetrahydroisoquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.