missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-1-benzofuran, 90%, Thermo Scientific™

Product Code. 10432400
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10432400 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10432400 Supplier Thermo Scientific Maybridge Supplier No. MAY00313DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 133720-60-2
Assay Percent Range 90%
Molecular Formula C8H5BrO
MDL Number MFCD09056756
Quantity 1 g
Synonym 7-bromobenzofuran, 7-bromobenzo b furan, benzofuran, 7-bromo, benzofuran, bromo, 7-bromo-benzofuran, 7-bromo-1-benzo b furan, acmc-1c18b
InChI Key GPZPQJITKRSVQJ-UHFFFAOYSA-N
SMILES BrC1=C2OC=CC2=CC=C1
IUPAC Name 7-bromo-1-benzofuran
Molecular Weight (g/mol) 197.03
PubChem CID 113628
Formula Weight 197.03
Chemical Name or Material 7-Bromo-1-benzofuran
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.