missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-1-hydroxyisoquinoline, 97%

Product Code. 11324775
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324775 5 g 5g
11314775 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11324775 Supplier Thermo Scientific Alfa Aesar Supplier No. L17800.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 223671-15-6 | C9H6BrNO | 224.057 g/mol

7-Bromo-1-hydroxyisoquinoline acts as a pharmaceutical intermediate. It is also used to prepare 1-amino-4-bromoisoquinoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Bromo-1-hydroxyisoquinoline acts as a pharmaceutical intermediate. It is also used to prepare 1-amino-4-bromoisoquinoline.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 223671-15-6
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.057
MDL Number MFCD02093963
InChI Key DSOKREQUHLPVFR-UHFFFAOYSA-N
PubChem CID 11276133
IUPAC Name 7-bromo-2H-isoquinolin-1-one
SMILES C1=CC(=CC2=C1C=CNC2=O)Br

Specifications

Melting Point 248°C to 250°C
Quantity 5 g
Beilstein 8405244
Synonym 7-bromoisoquinolin-1-ol, 7-bromo-1-hydroxyisoquinoline, 7-bromoisoquinolin-1 2h-one, 1 2h-isoquinolinone, 7-bromo, 1 2h-isoquinolinone,7-bromo, 7-bromo-1,2-dihydroisoquinolin-1-one, 7-bromoisoquinolone, 7-bromoisoquinolinone, 7-bromoisocarbostyril, 7-bromo-1-isoquinolinol
Solubility Information Slightly soluble in water.
Formula Weight 224.06
Percent Purity ≥98%
Chemical Name or Material 7-Bromo-1-hydroxyisoquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.