missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-1-methyl-3,4-dihydro-2(1H)-quinolinone, 96%

Product Code. 15408365
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408365 1 g 1g
15418365 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15408365 Supplier Thermo Scientific Alfa Aesar Supplier No. H33197.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1086386-20-0 | C10H10BrNO | 240.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1086386-20-0
Molecular Formula C10H10BrNO
Molecular Weight (g/mol) 240.10
MDL Number MFCD11655616
InChI Key XVBPEXCFWANWIM-UHFFFAOYSA-N
Synonym 7-bromo-1-methyl-3,4-dihydro-2 1h-quinolinone, 7-bromo-1-methyl-3,4-dihydroquinolin-2 1h-one, 2 1h-quinolinone, 7-bromo-3,4-dihydro-1-methyl
PubChem CID 46741415
IUPAC Name 7-bromo-1-methyl-3,4-dihydroquinolin-2-one
SMILES CN1C(=O)CCC2=C1C=C(Br)C=C2

Specifications

Melting Point 130°C to 134°C
Quantity 1 g
Formula Weight 240.1
Percent Purity 96%
Chemical Name or Material 7-Bromo-1-methyl-3,4-dihydro-2(1H)-quinolinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.