missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals

Product Code. 15407568
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15407568 1 g 1g
15417568 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15407568 Supplier Thermo Scientific Alfa Aesar Supplier No. H61071.03
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 105679-22-9 | Molecular Formula: C8H8BrNO | Molecular Weight: 214.062 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105679-22-9
Molecular Formula C8H8BrNO
Molecular Weight (g/mol) 214.062
MDL Number MFCD09056750
InChI Key JLZUUGCTPRPFKZ-UHFFFAOYSA-N
PubChem CID 18008960
IUPAC Name 7-bromo-3,4-dihydro-2H-1,4-benzoxazine
SMILES C1COC2=C(N1)C=CC(=C2)Br

Specifications

Quantity 1 g
Synonym 7-bromo-3,4-dihydro-2h-benzo b 1,4 oxazine, 7-bromo-3,4-dihydro-2h-benzo 1,4 oxazine, 2h-1,4-benzoxazine,7-bromo-3,4-dihydro, 2h-1,4-benzoxazine, 7-bromo-3,4-dihydro, pubchem22676, acmc-209yv4, 7-bromo-3,4-dihydro-2h-benzo b 1,4 oxazin
Formula Weight 214.06
Percent Purity 97%
Chemical Name or Material 7-Bromo-3,4-dihydro-2H-1,4-benzoxazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.