missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-4-methyl-3,4-dihydro-2H-1,4-benzoxazine, 95%, Thermo Scientific™

Product Code. 10732083
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10732083 1 g Amber Glass Bottle 1g
10662243 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10732083 Supplier Thermo Scientific Maybridge Supplier No. CC13510DA
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 154264-95-6
Molecular Formula C9H10BrNO
Molecular Weight (g/mol) 228.09
MDL Number MFCD02681913
InChI Key MQMFOFZKZBLSAB-UHFFFAOYSA-N
Synonym 7-bromo-4-methyl-3,4-dihydro-2h-1,4-benzoxazine, 7-bromo-3,4-dihydro-4-methyl-2h-1,4-benzoxazine, 2h-1,4-benzoxazine, 7-bromo-3,4-dihydro-4-methyl, 7-bromo-4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine, 7-bromo-4-methyl-2h,3h-benzo e 1,4-oxazine, 2h-1,4-benzoxazine,7-bromo-3,4-dihydro-4-methyl, 7-bromo-4-methyl-3,4-dihydro-2h-benzo 1,4 oxazine
PubChem CID 2776405
IUPAC Name 7-bromo-4-methyl-2,3-dihydro-1,4-benzoxazine
SMILES CN1CCOC2=C1C=CC(Br)=C2

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 228.09
Percent Purity 95%
Chemical Name or Material 7-Bromo-4-methyl-3, 4-dihydro-2H-1, 4-benzoxazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.