missing translation for 'onlineSavingsMsg'
Learn More

7-Bromoindole, 97%, Thermo Scientific™

Product Code. 10561824
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10561824 10 g 10g
10482895 1 g 1g
10001834 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10561824 Supplier Thermo Scientific Maybridge Supplier No. MO00800EA
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 51417-51-7
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.047
InChI Key RDSVSEFWZUWZHW-UHFFFAOYSA-N
Synonym 7-bromoindole, 1h-indole, 7-bromo, 7-bromo-indole, 7-bromindol, 7-bromoindol, 7-brom-1h-indole, pubchem7318, acmc-209ktm, ksc269i1f
PubChem CID 2757020
IUPAC Name 7-bromo-1H-indole
SMILES C1=CC2=C(C(=C1)Br)NC=C2

Specifications

Assay Percent Range 97%
Quantity 10 g
Formula Weight 196.05
Percent Purity 97%
Chemical Name or Material 7-Bromo-1H-indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.