missing translation for 'onlineSavingsMsg'
Learn More

7-Bromopyrrolo[2,1-f][1,2,4]triazin-4-amine, Thermo Scientific™

Product Code. 17372491 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 17372491

Brand: Thermo Scientific Acros 468700010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 937046-98-5
Molecular Formula C6H5BrN4
Molecular Weight (g/mol) 213.04
MDL Number MFCD09033848
InChI Key YGCJBESZJIGDMP-UHFFFAOYSA-N
IUPAC Name 7-bromopyrrolo[2,1-f][1,2,4]triazin-4-amine
SMILES NC1=NC=NN2C(Br)=CC=C12

Specifications

Infrared Spectrum Conforms to structure
Packaging Glass Bottle
Quantity 1 g
Formula Weight 213.04
Physical Form Powder
Chemical Name or Material 7-Bromopyrrolo[2,1-f][1,2,4]triazin-4-amine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.