missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Bromopyrrolo[2,1-f][1,2,4]triazin-4-amine, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Infrared Spectrum | Conforms to structure |
| Packaging | Glass Bottle |
| Quantity | 1 g |
| Formula Weight | 213.04 |
| Physical Form | Powder |
| Chemical Name or Material | 7-Bromopyrrolo[2,1-f][1,2,4]triazin-4-amine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction