missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-L-Tryptophan, TRC

Product Code. 30354969
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354969 25 mg -
30351539 2.5 mg 2.5mg
30349103 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30354969 Supplier Toronto Research Chemicals Supplier No. TRCC38615825MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73945-46-7
Molecular Formula C11H11N2O2Cl
Molecular Weight (g/mol) 238.67
IUPAC Name (2S)-2-amino-3-(7-chloro-1H-indol-3-yl)propanoic acid
SMILES N[C@@H](Cc1c[nH]c2c(Cl)cccc12)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C11H11ClN2O2/c12-8-3-1-2-7-6(5-14-10(7)8)4-9(13)11(15)16/h1-3,5,9,14H,4,13H2,(H,15,16)/t9-/m0/s1
Formula Weight 238.0509
Chemical Name or Material 7-Chloro-L-Tryptophan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.