missing translation for 'onlineSavingsMsg'
Learn More

7-Chlorobenzo[b]thiophene-3-methanol, TRC

Product Code. 30135203
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135203 250 mg 250mg
30132885 100 mg 100mg
30003949 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135203 Supplier Toronto Research Chemicals Supplier No. TRCC292850250MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142181-53-1
Molecular Formula C9 H7 Cl O S
Molecular Weight (g/mol) 198.6693
Synonym Benzo[b]thiophene-3-methanol, 7-chloro-, FI 7011, (7-chloro-1-benzothiophen-3-yl)methanol, Sertaconazole Imp. C, (7-Chloro-1-benzothiophen-3-yl)methanol
IUPAC Name (7-chloro-1-benzothiophen-3-yl)methanol
SMILES OCc1csc2c(Cl)cccc12

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H7ClOS/c10-8-3-1-2-7-6(4-11)5-12-9(7)8/h1-3,5,11H,4H2
Formula Weight 197.9906
Chemical Name or Material (7-Chloro-1-benzothiophen-3-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.