missing translation for 'onlineSavingsMsg'
Learn More

7-Chloroindole, 98%

Product Code. 11318586
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318586 250 mg 250mg
11308586 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318586 Supplier Thermo Scientific Alfa Aesar Supplier No. L17637.MD
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 53924-05-3 | C8H6ClN | 151.593 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53924-05-3
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
MDL Number MFCD01321301
InChI Key WMYQAKANKREQLM-UHFFFAOYSA-N
Synonym 7-chloroindole, 1h-indole, 7-chloro, 7-chloro indole, indole-7-chloro, 7-chlor-1h-indol, 7-chloro-indole, indole, 7-chloro, pubchem3195, acmc-1b1fq
PubChem CID 104644
IUPAC Name 7-chloro-1H-indole
SMILES C1=CC2=C(C(=C1)Cl)NC=C2

Specifications

Melting Point 57°C to 58°C
Quantity 250 mg
Beilstein 114743
Formula Weight 151.6
Percent Purity 98%
Chemical Name or Material 7-Chloroindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.