missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ 7-Chlorokynurenic acid

Product Code. 30001066
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001066 50 mg 50mg
30001065 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001066 Supplier R&D Systems™ Supplier No. 0237/50
  • ONLINE EXCLUSIVE PRICE
    502.35 €
    452.55 € / 50mg
    Save 49.80 €
    9% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent competitive inhibitor of L-glutamate uptake

7-Chlorokynurenic acid is an NMDA receptor antagonist acting at the glycine site. Potent competitive inhibitor of L-glutamate transport into synaptic vesicles.

Sodium Salt also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18000-24-3
Molecular Formula C10H6ClNO3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 223.62
Purity >98%
Molecular Weight (g/mol) 223.61
Chemical Name or Material 7-Chlorokynurenic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.