missing translation for 'onlineSavingsMsg'
Learn More

7-Chlorokynurenic acid, 98+%

Product Code. 15413969
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15413969 50 mg 50mg
15403969 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15413969 Supplier Thermo Scientific Alfa Aesar Supplier No. J61359.MF
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

NMDA receptor antagonist at the glycine site | CAS: 18000-24-3 | C10H6ClNO3 | 223.61 g/mol

NMDA receptor antagonist at the glycine site

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18000-24-3
Molecular Formula C10H6ClNO3
Molecular Weight (g/mol) 223.61
MDL Number MFCD00069227
InChI Key UAWVRVFHMOSAPU-UHFFFAOYSA-N
Synonym 7-chlorokynurenic acid, 7-chloro-4-hydroxyquinoline-2-carboxylic acid, 7-cl-kyna, 7-chloro-kynurenic acid, unii-s7936qon2k, 7-chloro-4-hydroxy-2-carboxyquinoline, 7-cka, 7-chloro-4-hydroxy-2-quinolinecarboxylic acid, 2-quinolinecarboxylic acid, 7-chloro-1,4-dihydro-4-oxo, cka
PubChem CID 1884
ChEBI CHEBI:63965
IUPAC Name 7-chloro-4-oxo-1,4-dihydroquinoline-2-carboxylic acid
SMILES OC(=O)C1=CC(=O)C2=CC=C(Cl)C=C2N1

Specifications

Color White to Yellow
Quantity 50 mg
Solubility Information Soluble in DMSO or dilute sodium hydroxide
Formula Weight 223.62
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material 7-Chlorokynurenic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.