missing translation for 'onlineSavingsMsg'
Learn More

7-Chlorokynurenic acid, MedChemExpress

Product Code. 16494456
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494456 100 mg 100mg
16449726 50 mg 50mg
16433178 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16494456 Supplier Medchem Express Supplier No. HY100811
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 7-Chlorokynurenic acid (7-CKA) is a potent and selective antagonist of the glycine B coagonist site of the N-methyl-D-aspartate (NMDA) receptor (IC50=0.56 μM). 7-Chlorokynurenic acid is also a potent inhibitor of the reuptake of glutamate into synaptic vesicles with a Ki of 0.59 μM. 7-Chlorokynurenic acid has potent antinociceptive actions after neuraxial delivery.

7-Chlorokynurenic acid (7-CKA) is a potent and selective antagonist of the glycine B coagonist site of the N-methyl-D-aspartate (NMDA) receptor (IC50=0.56 μM). 7-Chlorokynurenic acid is also a potent inhibitor of the reuptake of glutamate into synaptic vesicles with a Ki of 0.59 μM. 7-Chlorokynurenic acid has potent antinociceptive actions after neuraxial delivery.

Specifications

Chemical Name or Material 7-Chlorokynurenic acid
CAS 18000-24-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C10H6ClNO3
Quantity 100 mg
Synonym 7-CKA
Solubility Information DMSO : 16.67 mg/mL (74.55 mM; Need ultrasonic) ∣H2O : 1 mg/mL (4.47 mM; Need ultrasonic)
SMILES O=C(C1=NC2=CC(Cl)=CC=C2C(O)=C1)O
Molecular Weight (g/mol) 223.61
Formula Weight 223.61
Percent Purity 99.13%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.