missing translation for 'onlineSavingsMsg'
Learn More

7-Chlorokynurenic acid, TRC

Product Code. 30350347
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30350347 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350347 Supplier Toronto Research Chemicals Supplier No. TRCC36695825MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-Chlorokynurenic acid
CAS 18000-24-3
Molecular Formula C10H6ClNO3
Quantity 25 mg
InChI Formula InChI=1S/C10H6ClNO3/c11-5-1-2-6-7(3-5)12-8(10(14)15)4-9(6)13/h1-4H,(H,12,13)(H,14,15)
SMILES OC(=O)c1cc(O)c2ccc(Cl)cc2n1
IUPAC Name 7-chloro-4-hydroxyquinoline-2-carboxylic acid
Molecular Weight (g/mol) 223.61
Formula Weight 223.0036
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.