missing translation for 'onlineSavingsMsg'
Learn More

7-Dehydro Desmosterol-d6, TRC

Product Code. 30365823
Change view
Click to view available options
Quantity:
0.25 mg
1 mg
5 mg
Unit Size:
0.25mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365823 5 mg 5mg
30355006 1 mg 1mg
30364291 0.25 mg 0.25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30365823 Supplier Toronto Research Chemicals Supplier No. TRCD2295575MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C27 D6 H36 O
Molecular Weight (g/mol) 388.659
Synonym (3beta)-Cholesta-5,7,24-trien-3-ol-d6, Cholesta-5,7,24-trien-3beta-ol-d6, 7-Dehydrodesmosterol-d6, Cholesta-5,7,24(25)-trien-3beta-ol-d6, Cholesta-5,7,24-triene-3beta-ol-d6
IUPAC Name (3S,9S,10R,13R,14R,17R)-10,13-dimethyl-17-[(2R)-7,7,7-trideuterio-6-(trideuteriomethyl)hept-5-en-2-yl]-2,3,4,9,11,12,14,15,16,17-decahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES [2H]C([2H])([2H])C(=CCC[C@@H](C)[C@H]1CC[C@H]2C3=CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C([2H])([2H])[2H]

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C27H42O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h7,9-10,19,21,23-25,28H,6,8,11-17H2,1-5H3/t19-,21+,23-,24+,25+,26+,27-/m1/s1/i1D3,2D3
Formula Weight 388.361
Chemical Name or Material 7-Dehydro Desmosterol-d6
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.