missing translation for 'onlineSavingsMsg'
Learn More

7-Desmethoxy-7-fluoro Elvitegravir, TRC

Product Code. 30179234
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179234 25 mg 25mg
30179662 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179234 Supplier Toronto Research Chemicals Supplier No. TRCD29041025MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 869893-92-5
Molecular Formula C22H20ClF2NO4
Molecular Weight (g/mol) 435.85
Synonym 6-(3-Chloro-2-fluorobenzyl)-7-fluoro-1-[(S)-1-hydroxymethyl-2-methylpropyl]-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid, 6-[(3-Chloro-2-fluorophenyl)methyl]-7-fluoro-1,4-dihydro-1-[(1S)-1-(hydroxymethyl)-2-methylpropyl]-4-oxo-3-quinolinecarboxylic Acid;
IUPAC Name 6-[(3-chloro-2-fluorophenyl)methyl]-7-fluoro-1-[(2S)-1-hydroxy-3-methylbutan-2-yl]-4-oxoquinoline-3-carboxylic acid
SMILES CC(C)[C@@H](CO)N1C=C(C(=O)O)C(=O)c2cc(Cc3cccc(Cl)c3F)c(F)cc12

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H20ClF2NO4/c1-11(2)19(10-27)26-9-15(22(29)30)21(28)14-7-13(17(24)8-18(14)26)6-12-4-3-5-16(23)20(12)25/h3-5,7-9,11,19,27H,6,10H2,1-2H3,(H,29,30)/t19-/m1/s1
Formula Weight 435.1049
Chemical Name or Material 7-Desmethoxy-7-fluoro Elvitegravir
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.