missing translation for 'onlineSavingsMsg'
Learn More

7-Epi 10-Desacetyl Paclitaxel, TRC

Product Code. 30138296
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138296 25 mg 25mg
30008006 10 mg 10mg
30012788 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138296 Supplier Toronto Research Chemicals Supplier No. TRCE58350025MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78454-17-8
Molecular Formula C45 H49 N O13
Molecular Weight (g/mol) 811.87
Synonym Benzenepropanoic acid, β-(benzoylamino)-α-hydroxy-, (2aR,4R,4aS,6R,9S,11S,12S,12aR,12bS)-12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,6,11-trihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, (αR,βS)-, 7,11-Methano-1H-cyclodeca[3,4]benz[1,2-b]oxete, benzenepropanoic acid deriv., 10-Deacetyl-7-epi-paclitaxel, 10-Deacetyl-7-epi-taxol, 10-Deacetyl-7-epipaclitaxel, 10-Deacetyl-7-epitaxol, 10-Desacetyl-7-epipaclitaxel, 7-Epi-10-deacetyltaxol, 7-epi-10-Deacetylpaclitaxel, 7-epi-10-Deacetyltaxol, Ormosin V, Paclitaxel Imp H (EP)
IUPAC Name [(1S,2S,3R,4S,7R,9R,10S,12R,15S)-4-acetyloxy-15-[(2R,3S)-3-benzamido-2-hydroxy-3-phenylpropanoyl]oxy-1,9,12-trihydroxy-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.03,10.04,7]heptadec-13-en-2-yl] benzoate
SMILES CC(=O)O[C@@]12CO[C@@H]1C[C@@H](O)[C@]3(C)[C@@H]2[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](OC(=O)[C@H](O)[C@@H](NC(=O)c6ccccc6)c7ccccc7)C(=C([C@@H](O)C3=O)C5(C)C)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C45H49NO13/c1-24-29(57-41(54)35(50)33(26-15-9-6-10-16-26)46-39(52)27-17-11-7-12-18-27)22-45(55)38(58-40(53)28-19-13-8-14-20-28)36-43(5,37(51)34(49)32(24)42(45,3)4)30(48)21-31-44(36,23-56-31)59-25(2)47/h6-20,29-31,33-36,38,48-50,55H,21-23H2,1-5H3,(H,46,52)/t29-,30+,31+,33-,34+,35+,36-,38-,43+,44-,45+/m0/s1
Formula Weight 811.3204
Percent Purity >95%
Chemical Name or Material 7-epi-10-Deacetyltaxol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.