missing translation for 'onlineSavingsMsg'
Learn More

7-Epi-10-oxo Docetaxel, TRC

Product Code. 30137589
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137589 1 mg 1mg
30135201 5 mg 5mg
18051730 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137589 Supplier Toronto Research Chemicals Supplier No. TRCE5886101MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162784-72-7
Molecular Formula C43 H51 N O14
Molecular Weight (g/mol) 805.86
Synonym Benzenepropanoic acid, β-[[(1,1-dimethylethoxy)carbonyl]amino]-α-hydroxy-, (2aR,4R,4aS,9S,11S,12S,12aR,12bS)-12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5,6-dioxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, (αR,βS)-, Benzenepropanoic acid, β-[[(1,1-dimethylethoxy)carbonyl]amino]-α-hydroxy-, 12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5,6-dioxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, [2aR-[2aα,4α,4aβ,9α(αR*,βS*),11α,12α,12aα,12bα]]-, 4-epi-6-Oxodocetaxel, 5β,20-Epoxy-1,7α-dihydroxy-9,10-dioxotax-11-ene-2α,4,13α-triyl 4-acetate 2-benzoate 13-[(2R,3S)-3-[[(1,1-dimethylethoxy)carbonyl]amino]-2-hydroxy-3-phenylpropanoate, 10-Deoxy-10-oxo-7-epidocetaxel, Docetaxel Trihydrate Imp D (EP), (2aR,4R,4aS,9S,11S,12S,12aR,12bS)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-Dodecahydro-4,9,11,12,12b-pentahydroxy-4a,8,13,13-tetramethyl-7,11-methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5,6-dione 12b-acetate 12-benzoate 9-ester with (2R,3S)-N-tert-butoxycarbonyl-3-phenylisoserine, 7-Epi-10-oxo Docetaxel, Benzenepropanoic acid, β-[[(1,1-dimethylethoxy)carbonyl]amino]-α-hydroxy-, (2aR,4R,4aS,9S,11S,12S,12aR,12bS)-12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5,6-dioxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, (αR,βS)-, Benzenepropanoic acid, β-[[(1,1-dimethylethoxy)carbonyl]amino]-α-hydroxy-, 12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5,6-dioxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, [2aR-[2aα,4α,4aβ,9α(αR*,βS*),11α,12α,12aα,12bα]]-, 4-epi-6-Oxodocetaxel, 5β,20-Epoxy-1,7α-dihydroxy-9,10-dioxotax-11-ene-2α,4,13α-triyl 4-acetate 2-benzoate 13-[(2R,3S)-3-[[(1,1-dimethylethoxy)carbonyl]amino]-2-hydroxy-3-phenylpropanoate, 10-Deoxy-10-oxo-7-epidocetaxel, Docetaxel Trihydrate Imp D (EP), 4-epi-6-Oxodocetaxel, (2aR,4R,4aS,9S,11S,12S,12aR,12bS)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-Dodecahydro-4,9,11,12,12b-pentahydroxy-4a,8,13,13-tetramethyl-7,11-methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5,6-dione 12b-acetate 12-benzoate 9-ester with (2R,3S)-N-tert-butoxycarbonyl-3-phenylisoserine
IUPAC Name [(1S,2S,3R,4S,7R,9R,10S,15S)-4-acetyloxy-1,9-dihydroxy-15-[(2R,3S)-2-hydroxy-3-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoyl]oxy-10,14,17,17-tetramethyl-11,12-dioxo-6-oxatetracyclo[11.3.1.03,10.04,7]heptadec-13-en-2-yl] benzoate
SMILES CC(=O)O[C@@]12CO[C@@H]1C[C@@H](O)[C@]3(C)[C@@H]2[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](OC(=O)[C@H](O)[C@@H](NC(=O)OC(C)(C)C)c6ccccc6)C(=C(C(=O)C3=O)C5(C)C)C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C43H51NO14/c1-22-26(55-37(51)32(48)30(24-15-11-9-12-16-24)44-38(52)58-39(3,4)5)20-43(53)35(56-36(50)25-17-13-10-14-18-25)33-41(8,34(49)31(47)29(22)40(43,6)7)27(46)19-28-42(33,21-54-28)57-23(2)45/h9-18,26-28,30,32-33,35,46,48,53H,19-21H2,1-8H3,(H,44,52)/t26-,27+,28+,30-,32+,33-,35-,41+,42-,43+/m0/s1
Formula Weight 805.331
Percent Purity >95%
Chemical Name or Material 7-Epi-10-oxo-docetaxel (Docetaxel Impurity D)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.