missing translation for 'onlineSavingsMsg'
Learn More

7-Ethoxy-4-methylcoumarin, TRC

Product Code. 30377347
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377347 5 g -
30372099 2.5 g 2.5g
30381689 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30377347 Supplier Toronto Research Chemicals Supplier No. TRCE9375905G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-05-8
Molecular Formula C12 H12 O3
Molecular Weight (g/mol) 204.22
Synonym Coumarin, 7-ethoxy-4-methyl- (6CI,8CI), 7-Ethoxy-4-methyl-2H-1-benzopyran-2-one, 4-Methyl-7-ethoxycoumarin, 4-Methylumbelliferone ethyl ether, 7-Ethoxy-4-methyl-2H-chromen-2-one, 7-Ethoxy-4-methylcoumarin, NSC 60561, Maraniol
IUPAC Name 7-ethoxy-4-methylchromen-2-one
SMILES CCOc1ccc2C(=CC(=O)Oc2c1)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H12O3/c1-3-14-9-4-5-10-8(2)6-12(13)15-11(10)7-9/h4-7H,3H2,1-2H3
Formula Weight 204.0786
Chemical Name or Material 4-Methyl-7-Ethoxycoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.