missing translation for 'onlineSavingsMsg'
Learn More

7-Ethoxy Resorufin, TRC

Product Code. 30134266
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134266 25 mg 25mg
30133405 1 mg 1mg
30005929 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134266 Supplier Toronto Research Chemicals Supplier No. TRCE89287525MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5725-91-7
Molecular Formula C14 H11 N O3
Molecular Weight (g/mol) 241.24
Synonym 7-Ethoxyresorufin
IUPAC Name 7-ethoxyphenoxazin-3-one
SMILES CCOc1ccc2N=C3C=CC(=O)C=C3Oc2c1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H11NO3/c1-2-17-10-4-6-12-14(8-10)18-13-7-9(16)3-5-11(13)15-12/h3-8H,2H2,1H3
Formula Weight 241.0739
Percent Purity >95%
Chemical Name or Material 7-Ethoxyresorufin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.