missing translation for 'onlineSavingsMsg'
Learn More

7-Ethoxycoumarin, TRC

Product Code. 30388473
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388473 1 g
30379139 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30388473 Supplier Toronto Research Chemicals Supplier No. TRCE8917251G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31005-02-4
Molecular Formula C11 H10 O3
Molecular Weight (g/mol) 190.2
Synonym 2H-1-Benzopyran-2-one, 7-ethoxy- (9CI, ACI), 7-Ethoxy-2H-1-benzopyran-2-one (ACI), Coumarin, 7-ethoxy- (6CI, 8CI), 7-Ethoxycoumarin, Ethylumbelliferone
IUPAC Name 7-ethoxychromen-2-one
SMILES CCOc1ccc2C=CC(=O)Oc2c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C11H10O3/c1-2-13-9-5-3-8-4-6-11(12)14-10(8)7-9/h3-7H,2H2,1H3
Formula Weight 190.063
Chemical Name or Material 7-Ethoxycoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.