missing translation for 'onlineSavingsMsg'
Learn More

7-Ethylindole, 97%, Thermo Scientific™

Product Code. 10391364 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10391364 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10391364 Supplier Thermo Scientific Acros Supplier No. 271141000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22867-74-9
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.21
MDL Number MFCD00143514
InChI Key PIIZLMYXLGYWTN-UHFFFAOYSA-N
Synonym 7-ethylindole, 1h-indole, 7-ethyl, 7-ethyl indole, pubchem7338, acmc-1cfby, 7-ethylindole gc
PubChem CID 2724711
IUPAC Name 7-ethyl-1H-indole
SMILES CCC1=C2NC=CC2=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.05g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.6020 to 1.6040 (20°C, 589nm)
Quantity 100 g
Formula Weight 145.2
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 7-Ethylindole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.