missing translation for 'onlineSavingsMsg'
Learn More

7-Fluoro-2,3,4,9-tetrahydro-1H-beta-carboline, Thermo Scientific™

Product Code. 10442560
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10442560 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10442560 Supplier Thermo Scientific Maybridge Supplier No. MAY00133DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 177858-80-9
Packaging Amber Glass Bottle
Molecular Formula C11H11FN2
Quantity 1 g
Synonym 7-fluoro-2,3,4,9-tetrahydro-1h-beta-carboline, 7-fluoro-1,2,3,4-tetrahydro-9h-pyrido 3,4-b indole, 7-fluoro-1h,2h,3h,4h,9h-pyrido 3,4-b indole, 7-fluoro-2,3,4,9-tetrahydro-1h-pyrido 3,4-b indole, 7-fluoro-1,2,3,4-tetrahydrobeta-carboline, 1h-pyrido 3,4-b indole, 7-fluoro-2,3,4,9-tetrahydro
InChI Key OTGGOPSBNVEBDA-UHFFFAOYSA-N
SMILES C1CNCC2=C1C3=C(N2)C=C(C=C3)F
IUPAC Name 7-fluoro-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole
Molecular Weight (g/mol) 190.221
PubChem CID 15077550
Formula Weight 190.22
Percent Purity 95%
Chemical Name or Material 7-Fluoro-2,3,4,9-tetrahydro-1H-β-carboline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.