missing translation for 'onlineSavingsMsg'
Learn More

7-Fluorobenzo-2-oxa-1,3-diazole-4-sulfonic acid ammonium salt, MP Biomedicals™

Product Code. 11475511 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475511 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11475511 Supplier MP Biomedicals Supplier No. 0215808625
 Restricted Item
Estimated Shipment: 23-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sensitive and specific fluorescent labeling reagent for low molecular weight compounds and for macromolecules containing thiol groups

Chemical Identifiers

CAS 84806-27-9
Molecular Formula C6H6FN3O4S
Molecular Weight (g/mol) 235.189
MDL Number MFCD02682109
InChI Key JXLHNMVSKXFWAO-UHFFFAOYSA-N
Synonym sbd-f, 7-fluorobenzofurazan-4-sulfonic acid ammonium salt, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulphonate, azanium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluorobenzo c 1,2,5 oxadiazole-4-sulfonate, 7-fluorobenzo c 1,2,5-oxadiazole-4-sulfonic acid, azamethane, 2,1,3-benzoxadiazole-4-sulfonic acid, 7-fluoro-, ammonium salt 1:1
PubChem CID 2737780
IUPAC Name azanium;7-fluoro-2,1,3-benzoxadiazole-4-sulfonate
SMILES C1=C(C2=NON=C2C(=C1)S(=O)(=O)[O-])F.[NH4+]

Specifications

Quantity 25 mg
Formula Weight 235.2
Physical Form Powder
Chemical Name or Material 7-Fluorobenzo-2-oxa-1, 3-diazole-4-sulfonic acid ammonium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.