missing translation for 'onlineSavingsMsg'
Learn More

7-Fluorobenzofurazan-4-sulfonamide, 98%

Product Code. 11374629
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374629 25 mg 25mg
11435617 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374629 Supplier Thermo Scientific Alfa Aesar Supplier No. B23586.MB
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 91366-65-3 | C6H4FN3O3S | 217.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91366-65-3
Molecular Formula C6H4FN3O3S
Molecular Weight (g/mol) 217.17
MDL Number MFCD00036687
InChI Key XROXHZMRDABMHS-UHFFFAOYSA-N
Synonym abd-f, 4-aminosulfonyl-7-fluoro-2,1,3-benzoxadiazole, 7-fluorobenzofurazan-4-sulfonamide, 4-fluoro-7-sulfamoylbenzofurazan, 7-fluorobenzo c 1,2,5 oxadiazole-4-sulfonamide, 4-aminosulphonyl-7-fluoro-2,1,3-benzoxadiazole, 7-fluorobenzo c 1,2,5-oxadiazole-4-sulfonamide, pubchem23230, 4-fluoro-7-sulfamoylbenzofurazane, 4-fluoro-7-sulfoamoylbenzofurazan
PubChem CID 122067
IUPAC Name 7-fluoro-2,1,3-benzoxadiazole-4-sulfonamide
SMILES NS(=O)(=O)C1=CC=C(F)C2=NON=C12

Specifications

Melting Point 143°C to 147°C
Quantity 25 mg
Formula Weight 217.17
Percent Purity 98%
Chemical Name or Material 7-Fluorobenzofurazan-4-sulfonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.