missing translation for 'onlineSavingsMsg'
Learn More

7-Fluorobenzofurazan-4-sulfonic acid ammonium salt, 99%, Thermo Scientific Chemicals

Product Code. 11400397
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11400397 50 mg 50mg
11364629 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400397 Supplier Thermo Scientific Alfa Aesar Supplier No. B21767.MF
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 84806-27-9 | Molecular Formula: C6H6FN3O4S | Molecular Weight: 235.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84806-27-9
Molecular Formula C6H6FN3O4S
Molecular Weight (g/mol) 235.189
MDL Number MFCD02682109
InChI Key JXLHNMVSKXFWAO-UHFFFAOYSA-N
Synonym sbd-f, 7-fluorobenzofurazan-4-sulfonic acid ammonium salt, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulphonate, azanium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate, ammonium 7-fluorobenzo c 1,2,5 oxadiazole-4-sulfonate, 7-fluorobenzo c 1,2,5-oxadiazole-4-sulfonic acid, azamethane, 2,1,3-benzoxadiazole-4-sulfonic acid, 7-fluoro-, ammonium salt 1:1
PubChem CID 2737780
IUPAC Name azanium;7-fluoro-2,1,3-benzoxadiazole-4-sulfonate
SMILES C1=C(C2=NON=C2C(=C1)S(=O)(=O)[O-])F.[NH4+]

Specifications

Melting Point ∼285°C to 295°C (decomposition)
Quantity 50 mg
Sensitivity Light sensitive
Formula Weight 235.19
Percent Purity 99%
Chemical Name or Material 7-Fluorobenzofurazan-4-sulfonic acid ammonium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.