missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-6-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one hydrate(2:3), 99%, BLD Pharm™

Product Code. 31430916
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31430916 5 g
31417837 25 g
31425175 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31430916 Supplier BLD Pharm Supplier No. BD1275055g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 66778-17-4
Molecular Formula C30H38O21
MDL Number MFCD27981444
Quantity 5 g
SMILES O=C1C=CC2=C(O1)C=C(O)C(O[C@H]3[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O3)=C2.O=C4C=CC5=C(O4)C=C(O)C(O[C@H]6[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O6)=C5.[H]O[H].[H]O[H].[H]O[H]
Molecular Weight (g/mol) 734.61
Percent Purity 99%
Chemical Name or Material 7-Hydroxy-6-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one hydrate(2:3)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.