missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy Granisetron Hydrochloride, TRC

Product Code. 30010529
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010529 1 mg 1mg
30008558 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010529 Supplier Toronto Research Chemicals Supplier No. TRCH9426751MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133841-04-0
Molecular Formula C18 H24 N4 O2 . Cl H
Molecular Weight (g/mol) 364.87
Synonym 1H-Indazole-3-carboxamide, 7-hydroxy-1-methyl-N-(9-methyl-9-azabicyclo[3.3.1]non-3-yl)-, monohydrochloride, endo- (9CI), 9-Azabicyclo[3.3.1]nonane, 1H-indazole-3-carboxamide deriv., 7-Hydroxy-1-methyl-N-[(3-endo)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-1H-indazole-3-carboxamide hydrochloride, 7-Hydroxygranisetron hydrochloride, 7-Hydroxy granisetron hydrochloride
IUPAC Name 7-hydroxy-1-methyl-N-[(1R,5S)-9-methyl-9-azabicyclo[3.3.1]nonan-3-yl]indazole-3-carboxamide;hydrochloride
SMILES Cl.CN1[C@@H]2CCC[C@H]1C[C@H](C2)NC(=O)c3nn(C)c4c(O)cccc34

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H24N4O2.ClH/c1-21-12-5-3-6-13(21)10-11(9-12)19-18(24)16-14-7-4-8-15(23)17(14)22(2)20-16;/h4,7-8,11-13,23H,3,5-6,9-10H2,1-2H3,(H,19,24);1H/t11-,12+,13-;
Formula Weight 364.1666
Percent Purity >95%
Chemical Name or Material 7-Hydroxygranisetron hydrochloride
Grade HPLC
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.