missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy Quetiapine (1.0 mg/mL in Methanol), TRC

Product Code. 30179284
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179284 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30179284 Supplier Toronto Research Chemicals Supplier No. TRCKIT124510X1ML
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 139079-39-3
Molecular Formula C21 H25 N3 O3 S
Quantity 10 x 1 mL
Synonym 7-Hydroxyquetiapine, Dibenzo[b,f][1,4]thiazepin-7-ol, 11-[4-[2-(2-hydroxyethoxy)ethyl]-1-piperazinyl]-, 11-[4-[2-(2-Hydroxyethoxy)ethyl]-1-piperazinyl]dibenzo[b,f][1,4]thiazepin-7-ol, 7-Hydroxyquetiapine, ICI 214227
InChI Formula InChI=1S/C21H25N3O3S/c25-12-14-27-13-11-23-7-9-24(10-8-23)21-17-3-1-2-4-19(17)28-20-15-16(26)5-6-18(20)22-21/h1-6,15,25-26H,7-14H2
SMILES OCCOCCN1CCN(CC1)C2=Nc3ccc(O)cc3Sc4ccccc24
IUPAC Name 6-[4-[2-(2-hydroxyethoxy)ethyl]piperazin-1-yl]benzo[b][1,4]benzothiazepin-2-ol
Molecular Weight (g/mol) 399.51
Formula Weight 399.1617
Concentration or Composition (by Analyte or Components) 7-Hydroxyquetiapine 1 mg/mL
Chemical Name or Material 7-Hydroxyquetiapine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.