missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy Quetiapine, TRC

Product Code. 30179435
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179435 25 mg 25mg
30179212 1 mg 1mg
30179141 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179435 Supplier Toronto Research Chemicals Supplier No. TRCH95322525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139079-39-3
Molecular Formula C21 H25 N3 O3 S
Molecular Weight (g/mol) 399.51
Synonym 7-Hydroxyquetiapine, Dibenzo[b,f][1,4]thiazepin-7-ol, 11-[4-[2-(2-hydroxyethoxy)ethyl]-1-piperazinyl]-, 11-[4-[2-(2-Hydroxyethoxy)ethyl]-1-piperazinyl]dibenzo[b,f][1,4]thiazepin-7-ol, 7-Hydroxyquetiapine, ICI 214227
IUPAC Name 6-[4-[2-(2-hydroxyethoxy)ethyl]piperazin-1-yl]benzo[b][1,4]benzothiazepin-2-ol
SMILES OCCOCCN1CCN(CC1)C2=Nc3ccc(O)cc3Sc4ccccc24

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H25N3O3S/c25-12-14-27-13-11-23-7-9-24(10-8-23)21-17-3-1-2-4-19(17)28-20-15-16(26)5-6-18(20)22-21/h1-6,15,25-26H,7-14H2
Formula Weight 399.1617
Percent Purity >95%
Chemical Name or Material 7-Hydroxyquetiapine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.