missing translation for 'onlineSavingsMsg'
Learn More

7-Iodo-3,4-dihydro-2H-1,5-benzodioxepine, 97%, Thermo Scientific™

Product Code. 10600466
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10600466 5 g 5g
10417042 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10600466 Supplier Thermo Scientific Maybridge Supplier No. CC13211DE
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 306934-90-7
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.073
MDL Number MFCD02677706
InChI Key CIZHAPZOOAJDCN-UHFFFAOYSA-N
Synonym 7-iodo-3,4-dihydro-2h-benzo b 1,4 dioxepine, 2h-1,5-benzodioxepin,3,4-dihydro-7-iodo
PubChem CID 2776396
IUPAC Name 7-iodo-3,4-dihydro-2H-1,5-benzodioxepine
SMILES C1COC2=C(C=C(C=C2)I)OC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 5 g
Formula Weight 276.07
Percent Purity 95%
Chemical Name or Material 7-Iodo-3, 4-dihydro-2H-1, 5-benzodioxepine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.