missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline (>90%), TRC

Product Code. 30469716
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30469716 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30469716 Supplier Toronto Research Chemicals Supplier No. TRCM27110210G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material 7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline
CAS 1810-74-8
Molecular Formula C13 H17 N O
Quantity 10 g
Synonym 1,2-Dihydro-7-methoxy-2,2,4-trimethylquinoline, 2,2,4-Trimethyl-7-methoxy-1,2-dihydroquinoline
InChI Formula InChI=1S/C13H17NO/c1-9-8-13(2,3)14-12-7-10(15-4)5-6-11(9)12/h5-8,14H,1-4H3
SMILES COc1ccc2C(=CC(C)(C)Nc2c1)C
IUPAC Name 7-methoxy-2,2,4-trimethyl-1H-quinoline
Molecular Weight (g/mol) 203.28
Formula Weight 203.131
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.