missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxy-3,4,5,6-tetrahydro-2H-azepine, 97%, Thermo Scientific™

Product Code. 10513883
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10513883 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10513883 Supplier Thermo Scientific Maybridge Supplier No. TL00603EA
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 2525-16-8
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H13NO
MDL Number MFCD00006938
Quantity 10 g
Synonym o-methylcaprolactim, caprolactim methyl ether, 2h-azepine, 3,4,5,6-tetrahydro-7-methoxy, 2-methoxy-1-azacycloheptene, o-methylhexanolactim, caprolactim-o-methyl ether, o-methylcaprolactam, 2-methoxy-4,5,6,7-tetrahydro-3h-azepine, 1-aza-2-methoxy-1-cycloheptene, .epsilon.-caprolactim ether
InChI Key DNXIQMQGKSQHPC-UHFFFAOYSA-N
SMILES COC1=NCCCCC1
IUPAC Name 7-methoxy-3,4,5,6-tetrahydro-2H-azepine
Molecular Weight (g/mol) 127.187
PubChem CID 75657
Formula Weight 127.19
Percent Purity 97%
Chemical Name or Material 7-Methoxy-3,4,5,6-tetrahydro-2H-azepine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.