missing translation for 'onlineSavingsMsg'
Learn More

7-Methylindole-3-carboxaldehyde, 98%

Product Code. 11368476
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11368476 5 g 5g
11378476 250 mg 250mg
11358476 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11368476 Supplier Thermo Scientific Alfa Aesar Supplier No. L17519.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4771-50-0 | C10H9NO | 159.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4771-50-0
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00047170
InChI Key KTUFZHVVJBHGKZ-UHFFFAOYSA-N
Synonym 7-methylindole-3-carboxaldehyde, 7-methylindole-3-carboxyaldehyde, 3-formyl-7-methylindole, 7-methylindole 3-carboxaldehyde, 1h-indole-3-carboxaldehyde, 7-methyl, 3-carboxaldehyde, 7-methylindole, 7-methyl-1h-indole-3-carboxaldehyde, 7-methylindole-3-carbaldehyde, pubchem8327, acmc-1ak55
PubChem CID 260389
IUPAC Name 7-methyl-1H-indole-3-carbaldehyde
SMILES CC1=C2NC=C(C=O)C2=CC=C1

Specifications

Melting Point 209°C to 211°C
Quantity 5 g
Beilstein 122340
Sensitivity Air sensitive
Formula Weight 159.19
Percent Purity 98%
Chemical Name or Material 7-Methylindole-3-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.