missing translation for 'onlineSavingsMsg'
Learn More

7-Methyluric Acid, TRC

Product Code. 30483623
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483623 5 mg 5mg
30476963 10 mg 10mg
30470320 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30483623 Supplier Toronto Research Chemicals Supplier No. TRCM3381855MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-37-3
Molecular Formula C6 H6 N4 O3
Molecular Weight (g/mol) 182.14
IUPAC Name 7-methyl-3,9-dihydropurine-2,6,8-trione
SMILES CN1C(=O)NC2=C1C(=O)NC(=O)N2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C6H6N4O3/c1-10-2-3(8-6(10)13)7-5(12)9-4(2)11/h1H3,(H3,7,8,9,11,12,13)
Formula Weight 182.044
Chemical Name or Material 7-Methyluric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.