missing translation for 'onlineSavingsMsg'
Learn More

7-Methylxanthine, 98%

Product Code. 15433847
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15433847 250 mg 250mg
11412260 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15433847 Supplier Thermo Scientific Alfa Aesar Supplier No. H29190.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 552-62-5 | C6H6N4O2 | 166.14 g/mol

It is used to metabolite theophylline and caffeine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to metabolite theophylline and caffeine.

Solubility
Soluble in dimethyl sulfoxide. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-62-5
Molecular Formula C6H6N4O2
Molecular Weight (g/mol) 166.14
MDL Number MFCD00037979
InChI Key PFWLFWPASULGAN-UHFFFAOYSA-N
Synonym 7-methylxanthine, heteroxanthine, heteroxanthin, 7-methylxanthin, 7-methyl-1h-purine-2,6 3h,7h-dione, 2,6-dihydroxy-7-methylpurine, xanthine, 7-methyl, 3,7-dihydro-7-methyl-1h-purine-2,6-dione, 7-methyl-3,7-dihydro-1h-purine-2,6-dione, 1h-purine-2,6-dione, 3,7-dihydro-7-methyl
PubChem CID 68374
ChEBI CHEBI:48991
SMILES CN1C=NC2=C1C(=O)NC(=O)N2

Specifikationer

Melting Point >300°C
Quantity 250 mg
Solubility Information Soluble in dimethyl sulfoxide. Insoluble in water.
IUPAC Name 7-methyl-3H-purine-2,6-dione
Formula Weight 166.14
Percent Purity 98%
Chemical Name or Material 7-Methylxanthine

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.