missing translation for 'onlineSavingsMsg'
Learn More

7-Oxabicyclo[2.2.1]heptane, 98%

Product Code. 11445207
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445207 100 g 100g
11425207 5 g 5g
11435207 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11445207 Supplier Thermo Scientific Alfa Aesar Supplier No. B22612.22
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 279-49-2 | C6H10O | 98.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 279-49-2
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00082342
InChI Key YPWFNLSXQIGJCK-UHFFFAOYSA-N
Synonym 1,4-epoxycyclohexane, 7-oxabicyclo 2.2.1 heptane, 7-oxanorbornane, 1,4-oxycyclohexane, 3,6-endoxocyclohexane, 3,6-endooxycyclohexane, cyclohexane,4-epoxy, cyclohexane,6-endoxo, cyclohexane, 1,4-epoxy
PubChem CID 9234
IUPAC Name 7-oxabicyclo[2.2.1]heptane
SMILES C1CC2CCC1O2

Specifications

Density 0.968
Quantity 100 g
Refractive Index 1.448
UN Number UN3271
Boiling Point 118°C to 120°C (713 mmHg)
Formula Weight 98.15
Percent Purity 98%
Chemical Name or Material 7-Oxabicyclo[2.2.1]heptane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.