missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 7300 System SDS v1.4 Upgrade Kit with RQ Study

Product Code. 15813384 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15813384 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15813384 Supplier Applied Biosystems™ Supplier No. 4379635

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This software includes an upgrade for the 7300 SDS Software to provide the capability and reliability of the most recent version.

Powerful and user-friendly, the 7300 Sequence Detection Software includes:
• Simple plate setup wizard is streamlined for each application – option to setup before or after the completing the run
• Correct plate-setup mistakes after the run without losing collected data
• One-click graphical export to PowerPoint or JPG for top notch presentations
• Flexible system can be used as a plate reader or a regular thermal cycler if needed
• Real-time amplification monitoring and flexibility to add additional PCR cycles during a run
• Lamp-lifetime monitoring ensures highest possible instrument performance

Additionally, the 7300 System SDS RQ Study Software, a gene expression analysis software package with powerful data viewing capabilities, is included. Data from up to 10 96-well RQ plates can be simultaneously analyzed. In an RQ study, you can:
• Select the endogenous control and the calibrator sample.
• Select the control type when applicable.
• Set baseline and threshold values and RQ Min/Max
Confidence Levels.
• Omit individual wells or sample replicates.

The user is able to view post-run amplification in three different formats, while gene expression plots show expression levels of target samples relative to the calibrator.

For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature
For Use With (Equipment) 7300 System
Software Type Gene Expression Analysis
Product Type Software Upgrade Kit
For Use With (Application) Real Time PCR (qPCR)
Operating System Windows XP Pro (Service Pk. 2)
Quantity 1 kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.