Learn More
R&D Systems™ 740 Y-P
Product Code. 18776376
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity Unit Size
18776376 1 mg 1mg
1 options
Product Code. 18776376 Supplier R&D Systems™ Supplier No. 1983/1
  • ONLINE EXCLUSIVE PRICE
    466.54 €
    417.90 € / 1mg
    Save 48.64 €
    10% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
A Tocris product, part of R&D Systems. Cell-permeable PI 3-kinase activator
740 Y-P is a cell-permeable phosphopeptide activator of PI 3-kinase. Binds with high affinity to p85 subunit of the enzyme. Displays mitogenic activity in C2 muscle cells and promotes survival of rat cerebellar granule neurons in vitro.
TRUSTED_SUSTAINABILITY
Specifications
Product Line Tocris
Quantity 1 mg