Learn More
Applied Biosystems™ 7500 Fast Real-Time PCR Systems Spectral Calibration Kit II
Shop All Thermo Scientific ProductsDescription
This easy-to-use kit establishes the pure dye spectra and multicomponent values for Cy3™, Cy5™, and Texas Red™ dyes on the 7500 Fast Real-Time PCR System. The Applied Biosystems™ 7500 Real-Time PCR Systems Spectral Calibration Kit II contains three Optical 96-well Fast Thermocycling Plates preloaded and sealed with Cy3™, Cy5™, and Texas Red™ fluorescent dye standards. Spend Your Time on Research, Not Instrument Calibration: This kit's easy-to-use, preloaded, 96-well optical reaction plates enable quick and easy calibration of the 7500 Fast Real-Time PCR System. Insert the reaction plates during system installation, and the system collects the dye standard's spectral information. The system's application algorithm then uses the calibration information to ensure accurate data analysis.
Kit II
- 96-well optical reaction plates preloaded with Cy3™, Cy5™, and Texas Red™ dyes allow quick and easy calibration of the 7500 Fast Real-Time PCR System
- Easy-to-use, preloaded spectral calibration kit allows accurate discrimination of the individual dyes in a reaction mixture when all the dyes are excited simultaneously
Kit l
- 96-well optical reaction plates preloaded with dyes (FAM™/SYBR™ Green I, VIC™/JOE™, NED™/TAMRA™/ROX™) allow quick and easy calibration of the 7500 Fast Real-Time PCR System
- Easy-to-use, preloaded spectral calibration kit allows accurate discrimination of the individual dyes in a reaction mixture when all the dyes are excited simultaneously
- Includes the Passive Reference I dye standard required for signal normalization in all real-time and endpoint reactions
For Research Use Only. Not for use in diagnostic procedures.
Order Info
Expiration date: 12 months from the date of delivery.
Specifications
Specifications
| Content And Storage | Contains three optical 96-Well Fast Thermocycling Plates preloaded and sealed, containing Cy3™ Texas Red™ and Cy5™ fluorescent dye standards. Store at -15 to -25°C. Protect from light. Expiration date: 12 months from the date of delivery |
| Calibrated Dye | Cy™3, Cy™5, Texas Red™ |
| For Use With (Equipment) | 7500 Fast System |
| Detection Method | Fluorescence |
| Product Type | Spectral Calibration Kit |
| For Use With (Application) | Real Time PCR (qPCR) |
| Shipping Condition | Dry Ice |
| Quantity | 1 kit |
| Calibration Type | Spectral Calibration |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.