missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 7900HT TaqMan™ Array Upgrade

Product Code. 15883354 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15883354 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15883354 Supplier Applied Biosystems™ Supplier No. 4329012
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The 7900HT TaqMan™ Array Upgrade enables you to run the TaqMan™ Arrays – a microfluidic card – on your Applied Biosystems 7900HT Fast Real-Time PCR System.

This upgrade gives you access to an efficient, easy-to-use tool that eliminates labor-intensive pipetting steps and enables high throughput gene expression results using the gold-standard 5' nuclease TaqMan™ Assay technology.

• Convenience – includes everything you need to run the 7900HT TaqMan™ Array on your 7900HT system
• Success – highly trained service engineers will install and test your 7900HT TaqMan™ Array Upgrade to get you up and running quickly
• High throughput – provides access to 384-well format without liquid-handling robotics

This upgrade to your 7900HT system provides everything you need to run the TaqMan™ Array.

For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY

Specifications

Block Format Interchangeable
Format 384-well Microfluidic Card
Content And Storage This kit contains a thermal cycling block, a microfluidic card sealer, centrifuge buckets with adapters, and a chemistry installation kit.
Reaction Speed Fast
Product Line TaqMan
Product Type Array Upgrade
For Use With (Application) Real Time PCR (qPCR)
Quantity 1 kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.