missing translation for 'onlineSavingsMsg'
Learn More

7ACC1, MedChemExpress

Product Code. 16438886
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438886 100 mg 100mg
16411476 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16438886 Supplier Medchem Express Supplier No. HYD0067
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 7ACC1(DEAC; Coumarin D 1421; D 1421) selectively interfere with lactate fluxes in the lactate-rich tumor microenvironment; inhibits lactate influx but not efflux in tumor cells expressing MCT1 and MCT4 transporters.

7ACC1(DEAC

Specifications

Chemical Name or Material 7ACC1
CAS 50995-74-9
Color Orange
Purity Grade Notes Research
Molecular Formula C14H15NO4
Quantity 100 mg
Synonym DEAC Coumarin D 1421 D 1421
Solubility Information DMSO : 33.33 mg/mL (127.57 mM; Need ultrasonic)
SMILES O=C(C1=CC2=CC=C(N(CC)CC)C=C2OC1=O)O
Molecular Weight (g/mol) 261.27
Formula Weight 261.27
Percent Purity 99.96%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.