missing translation for 'onlineSavingsMsg'
Learn More

7beta-Hydroxy Dehydro Epiandrosterone, TRC

Product Code. 30391396
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30391396 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391396 Supplier Toronto Research Chemicals Supplier No. TRCH825206250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7b-Hydroxy DHEA
CAS 2487-48-1
Molecular Formula C19 H28 O3
Quantity 250 mg
Synonym Androst-5-en-17-one, 3,7-dihydroxy-, (3β,7β)-, Androst-5-en-17-one, 3β,7β-dihydroxy- (7CI,8CI), (3β,7β)-3,7-Dihydroxyandrost-5-en-17-one, 3α,7β-Hydroxyandrost-5-en-17-one, 3β,7β-Dihydroxy-5-androsten-17-one, 3β,7β-Dihydroxy-5-androstene-17-one, 7-β-OH-DHEA, 7β-Hydroxy-DHEA, 7β-Hydroxydehydroepiandrosterone, Androst-5-en-17-one-3β,7β-diol, Androst-5-ene-17-one-3β,7β-diol, Androst-5-ene-3β,7β-diol-17-one
InChI Formula InChI=1S/C19H28O3/c1-18-7-5-12(20)9-11(18)10-15(21)17-13-3-4-16(22)19(13,2)8-6-14(17)18/h10,12-15,17,20-21H,3-9H2,1-2H3/t12-,13-,14-,15-,17-,18-,19-/m0/s1
SMILES C[C@]12CC[C@H]3[C@@H]([C@@H](O)C=C4C[C@@H](O)CC[C@]34C)[C@@H]1CCC2=O
IUPAC Name (3S,7R,8R,9S,10R,13S,14S)-3,7-dihydroxy-10,13-dimethyl-1,2,3,4,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-one
Molecular Weight (g/mol) 304.42
Formula Weight 304.2038
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.